2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 98%

Supplier: Ambeed
Warning

A942976-100MG A942976-1G A942976-250MG A942976-5G
76833-006EA 37.14 USD
76833-006 76833-008 76833-010 76833-012
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide 98%
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

Precaución: for Research Use Only

Formula: C₁₃H₁₇BClNO₃
MW: 281.54 g/mol
MDL Number: MFCD12546509
CAS Number: 1218791-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR