7-Chloro-1-(2-methyl-4-nitrobenzoyl)-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one 98% (by HPLC)

Supplier: Ambeed
Warning

A942958-1G A942958-25G A942958-5G
76833-000EA 18 USD
76833-000 76833-002 76833-004
7-Chloro-1-(2-methyl-4-nitrobenzoyl)-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one 98% (by HPLC)
7-Chloro-1-(2-methyl-4-nitrobenzoyl)-3,4-dihydro-1H-benzo[b]azepin-5(2H)-one

Precaución: for Research Use Only

Formula: C₁₈H₁₅ClN₂O₄
MDL Number: MFCD19440800
CAS Number: 137982-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR