4-Bromo-1,2-bis(bromomethyl)benzene 97%

Supplier: Ambeed
Danger

A940641-1G A940641-250MG A940641-25G A940641-5G A940641-10G
76832-986EA 79.57 USD
76832-986 76832-988 76832-990 76832-992 77614-656
4-Bromo-1,2-bis(bromomethyl)benzene 97%
4-Bromo-1,2-bis(bromomethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₇Br₃
MW: 342.85 g/mol
MDL Number: MFCD16620247
CAS Number: 69189-19-1
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR