1-(Methylsulfonyl)piperidin-4-ol 98%

Supplier: Ambeed
Warning

A933719-100MG A933719-1G A933719-250MG
76832-966EA 15 USD
76832-966 76832-968 76832-970
1-(Methylsulfonyl)piperidin-4-ol 98%
1-(Methylsulfonyl)piperidin-4-ol

Precaución: for Research Use Only

Formula: C₆H₁₃NO₃S
MDL Number: MFCD11044218
CAS Number: 141482-19-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR