2-(2-Ethoxyphenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one 95%

Supplier: Ambeed
Warning

A933025-1G A933025-250MG A933025-25G A933025-5G
76832-958EA 19 USD
76832-958 76832-960 76832-962 76832-964
2-(2-Ethoxyphenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one 95%
2-(2-Ethoxyphenyl)-5-methyl-7-propylimidazo[5,1-f][1,2,4]triazin-4(3H)-one

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₄O₂
MW: 312.36 g/mol
MDL Number: MFCD09833131
CAS Number: 224789-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR