N,N-Dimethyl-1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine 95%

Supplier: Ambeed
Warning

Synonyms: N,N-Dimethyl-1-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanamine

A929818-1G A929818-250MG A929818-5G
76832-944EA 68.96 USD
76832-944 76832-946 76832-948
N,N-Dimethyl-1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine 95%
N,N-Dimethyl-1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methylamine

Precaución: for Research Use Only

Formula: C₁₅H₂₄BNO₂
MW: 261.168 g/mol
MDL Number: MFCD08669568
CAS Number: 909391-56-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR