3-Chloro-5-propoxyphenylboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-(3-Chloro-5-propoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A926347-100MG A926347-1G A926347-250MG
76832-932EA 36.08 USD
76832-932 76832-934 76832-936
3-Chloro-5-propoxyphenylboronic acid pinacol ester 98%
3-Chloro-5-propoxyphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BClO₃
MW: 296.6000 g/mol
MDL Number: MFCD12546578
CAS Number: 1218789-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR