2-Formyl-4-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

A922475-1G A922475-250MG A922475-100MG A922475-5G
76832-922EA 80 USD
76832-922 76832-924 77886-911 77886-912
2-Formyl-4-(trifluoromethyl)phenylboronic acid 98%
2-Formyl-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₆BF₃O₃
MDL Number: MFCD13195656
CAS Number: 1217501-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR