4-(2-Methoxyethoxy)benzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-(2-Methoxyethoxy)phenyl)boronic acid

A920211-10G A920211-1G A920211-5G A920211-250MG A920211-25G
76832-914EA 78 USD
76832-914 76832-916 76832-918 77886-909 77886-910
4-(2-Methoxyethoxy)benzeneboronic acid 97%
4-(2-Methoxyethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BO₄
MW: 196.009 g/mol
MDL Number: MFCD03452784
CAS Number: 265664-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR