5-Bromo-2-(trimethylsilyl)pyridine 95+%

Supplier: Ambeed
Warning

A918284-1G A918284-250MG
76832-910EA 173.99 USD
76832-910 76832-912
5-Bromo-2-(trimethylsilyl)pyridine 95+%
5-Bromo-2-(trimethylsilyl)pyridine

Precaución: for Research Use Only

Formula: C₈H₁₂BrNSi
MDL Number: MFCD18839255
CAS Number: 291312-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR