2,5-Dichloro-3-fluorobenzoic acid 95+%

Supplier: Ambeed
Warning

A914697-1G A914697-250MG A914697-5G
76832-898EA 31.83 USD
76832-898 76832-900 76832-902
2,5-Dichloro-3-fluorobenzoic acid 95+%
2,5-Dichloro-3-fluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂FO₂
MW: 209.002 g/mol
MDL Number: MFCD13250174
CAS Number: 501008-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR