N,N,N-Trimethyloctadecan-1-aminium iodide 95%

Supplier: Ambeed
Warning

A911389-250MG A911389-5G
76832-894EA 29 USD
76832-894 76832-896
N,N,N-Trimethyloctadecan-1-aminium iodide 95%
N,N,N-Trimethyloctadecan-1-aminium iodide

Precaución: for Research Use Only

Formula: C₂₁H₄₆IN
MDL Number: MFCD00959638
CAS Number: 4292-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR