4,4'-Azanediylbis(butan-1-ol) 95%

Supplier: Ambeed
Warning

A909407-100MG A909407-1G A909407-250MG A909407-5G
76832-886EA 38.2 USD
76832-886 76832-888 76832-890 76832-892
4,4'-Azanediylbis(butan-1-ol) 95%
4,4'-Azanediylbis(butan-1-ol)

Precaución: for Research Use Only

Formula: C₈H₁₉NO₂
MW: 161.24 g/mol
MDL Number: MFCD20661326
CAS Number: 79448-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR