Methyl 2,6-dimethylbenzoate 98%

Supplier: Ambeed
Warning

A908468-1G A908468-250MG A908468-5G
76832-882EA 54.11 USD
76832-882 76832-884 77684-865
Methyl 2,6-dimethylbenzoate 98%
Methyl 2,6-dimethylbenzoate

Precaución: For research use only.

Formula: C₁₀H₁₂O₂
MW: 164.201 g/mol
MDL Number: MFCD06203721
CAS Number: 14920-81-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR