4-(Hydrazinocarbonyl)benzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Hydrazinecarbonyl)phenyl)boronic acid

A907911-1G A907911-250MG A907911-5G
76832-878EA 71.09 USD
76832-878 76832-880 77684-864
4-(Hydrazinocarbonyl)benzeneboronic acid 98%
4-(Hydrazinocarbonyl)benzeneboronic acid

Precaución: For research use only.

Formula: C₇H₉BN₂O₃
MW: 179.97 g/mol
MDL Number: MFCD06659865
CAS Number: 850567-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR