5-Iodo-2-nitrophenol 98%

Supplier: Ambeed
Danger

A906978-100MG A906978-1G A906978-250MG A906978-5G
76832-872EA 15 USD
76832-872 76832-874 76832-876 77886-906
5-Iodo-2-nitrophenol 98%
5-Iodo-2-nitrophenol

Precaución: for Research Use Only

Formula: C₆H₄INO₃
MDL Number: MFCD12923199
CAS Number: 27783-55-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR