4,4,5,5-Tetramethyl-2-(3-methyl-4-nitrophenyl)-1,3,2-dioxaborolane 95+%

Supplier: Ambeed
Warning

A905342-100MG A905342-1G A905342-250MG A905342-5G
76832-866EA 36 USD
76832-866 76832-868 76832-870 77886-905
4,4,5,5-Tetramethyl-2-(3-methyl-4-nitrophenyl)-1,3,2-dioxaborolane 95+%
4,4,5,5-Tetramethyl-2-(3-methyl-4-nitrophenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₄
MDL Number: MFCD10699700
CAS Number: 590418-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR