3-(2-(4-(o-Tolyl)piperazin-1-yl)ethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Dapiprazole hydrochloride

A893853-100MG A893853-10MG A893853-1MG A893853-25MG A893853-50MG A893853-5MG
76832-834EA 122.01 USD
76832-834 76832-836 76832-838 76832-840 76832-842 76832-844
3-(2-(4-(o-Tolyl)piperazin-1-yl)ethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine hydrochloride 98%
3-(2-(4-(o-Tolyl)piperazin-1-yl)ethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyridine hydrochloride

Precaución: for Research Use Only

Formula: C₁₉H₂₈ClN₅
MW: 361.91 g/mol
MDL Number: MFCD00941400
CAS Number: 72822-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR