4-Bromo-1-ethoxy-2-fluorobenzene 97%

Supplier: Ambeed
Warning

A893765-1G A893765-5G
76832-830EA 22 USD
76832-830 76832-832
4-Bromo-1-ethoxy-2-fluorobenzene 97%
4-Bromo-1-ethoxy-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
MDL Number: MFCD09907659
CAS Number: 115467-08-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR