6-Chloroquinoline-2-carbonitrile 97%

Supplier: Ambeed
Warning

A892770-1G A892770-250MG
76832-826EA 74 USD
76832-826 76832-828
6-Chloroquinoline-2-carbonitrile 97%
6-Chloroquinoline-2-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₅ClN₂
MDL Number: MFCD17926278
CAS Number: 52313-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR