Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-methylimidazo[1,2-a]pyridine-2-carboxylate

A892325-100MG A892325-1G A892325-250MG A892325-5G
76832-818EA 26.53 USD
76832-818 76832-820 76832-822 76832-824
Ethyl-6-methylimidazo[1,2-a]pyridine-2-carboxylate 98%
Ethyl 6-methylimidazo[1,2-a]pyridine-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂O₂
MW: 204.2300 g/mol
MDL Number: MFCD02325396
CAS Number: 70705-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR