4-Chloro-1H-indazole-6-carboxylic acid 97%

Supplier: Ambeed
Warning

A891557-100MG A891557-250MG A891557-5G
76832-812EA 25.47 USD
76832-812 76832-814 76832-816
4-Chloro-1H-indazole-6-carboxylic acid 97%
4-Chloro-1H-indazole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MW: 196.59 g/mol
MDL Number: MFCD07781794
CAS Number: 885523-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR