3-Methacrylamido-N,N,N-trimethylpropan-1-aminium chloride 50% (w/w) in water

Supplier: Ambeed
Warning

A890446-1000G A890446-100G A890446-25G A890446-500G
76832-804EA 202 USD
76832-804 76832-806 76832-808 76832-810
3-Methacrylamido-N,N,N-trimethylpropan-1-aminium chloride 50% (w/w) in water
3-Methacrylamido-N,N,N-trimethylpropan-1-aminium chloride

Precaución: for Research Use Only

Formula: C₁₀H₂₁ClN₂O
MDL Number: MFCD00011812
CAS Number: 51410-72-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR