6-Bromo-N-methylquinolin-4-amine 95%

Supplier: Ambeed
Warning

A890440-100MG A890440-1G A890440-250MG
76832-798EA 85.94 USD
76832-798 76832-800 76832-802
6-Bromo-N-methylquinolin-4-amine 95%
6-Bromo-N-methylquinolin-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂
MW: 237.10 g/mol
MDL Number: MFCD14705170
CAS Number: 916812-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR