(R)-tert-Butyl (2-oxoazepan-3-yl)carbamate 95%

Supplier: Ambeed
Warning

A890312-100MG A890312-250MG
76832-794EA 65 USD
76832-794 76832-796
(R)-tert-Butyl (2-oxoazepan-3-yl)carbamate 95%
(R)-tert-Butyl (2-oxoazepan-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₀N₂O₃
MDL Number: MFCD03788637
CAS Number: 106691-72-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR