5-Cyano-2-(trifluoromethoxy)benzeneboronic acid 98%

Supplier: Ambeed
Warning

A888455-100MG A888455-1G A888455-250MG A888455-5G
76832-788EA 14 USD
76832-788 76832-790 76832-792 77886-887
5-Cyano-2-(trifluoromethoxy)benzeneboronic acid 98%
5-Cyano-2-(trifluoromethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₅BF₃NO₃
MDL Number: MFCD11053857
CAS Number: 1072946-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR