GW788388 98%

Supplier: Ambeed
Warning

Synonyms: 4-(4-(3-(Pyridin-2-yl)-1H-pyrazol-4-yl)pyridin-2-yl)-N-(tetrahydro-2H-pyran-4-yl)benzamide

A887665-10MG A887665-1MG A887665-50MG A887665-5MG
76832-780EA 55.17 USD
76832-780 76832-782 76832-784 76832-786
GW788388 98%
GW788388

Precaución: for Research Use Only

Formula: C₂₅H₂₃N₅O₂
MW: 425.4824 g/mol
MDL Number: MFCD16038936
CAS Number: 452342-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR