4-Fluoro-2-methyl-5-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Carboxy-2-fluoro-4-methylnitrobenzene, 2-Carboxy-5-fluoro-4-nitrotoluene

A885564-1G A885564-25G A885564-5G
76832-774EA 7 USD
76832-774 76832-776 76832-778
4-Fluoro-2-methyl-5-nitrobenzoic acid 98%
4-Fluoro-2-methyl-5-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₄
MDL Number: MFCD11847453
CAS Number: 64695-92-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR