2-(5-Bromo-2-chlorophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

A885386-1G A885386-250MG A885386-5G
76832-768EA 44.56 USD
76832-768 76832-770 76832-772
2-(5-Bromo-2-chlorophenyl)acetonitrile 98%
2-(5-Bromo-2-chlorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₅BrClN
MW: 230.49 g/mol
MDL Number: MFCD18390591
CAS Number: 1057216-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR