4-Methyl-4-phenylpentan-2-ol 95%

Supplier: Ambeed
Warning

A885102-100MG A885102-250MG
76832-764EA 201.58 USD
76832-764 76832-766
4-Methyl-4-phenylpentan-2-ol 95%
4-Methyl-4-phenylpentan-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₁₈O
MW: 178.27 g/mol
MDL Number: MFCD00087654
CAS Number: 2035-93-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR