3-Amino-4-iodobenzonitrile 98%

Supplier: Ambeed
Danger

A882776-1G A882776-5G
76832-752EA 103.97 USD
76832-752 76832-754
3-Amino-4-iodobenzonitrile 98%
3-Amino-4-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MW: 244.03 g/mol
MDL Number: MFCD18389612
CAS Number: 665033-21-6
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR