3-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 96%

Supplier: Ambeed
Warning

A881939-100MG A881939-1G A881939-250MG A881939-5G
76832-744EA 9 USD
76832-744 76832-746 76832-748 76832-750
3-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 96%
3-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₂
MDL Number: MFCD09260444
CAS Number: 624741-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR