2-Acetamido-5-pyridineboronic acid pinacol ester hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: N-(5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl)acetamide hydrochloride

A880840-100MG A880840-1G A880840-250MG
76832-738EA 37.14 USD
76832-738 76832-740 76832-742
2-Acetamido-5-pyridineboronic acid pinacol ester hydrochloride 97%
2-Acetamido-5-pyridineboronic acid pinacol ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₀BClN₂O₃
MW: 298.58 g/mol
MDL Number: MFCD12026097
CAS Number: 1150561-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR