2,4-Difluoro-5-iodobenzoic acid 95%

Supplier: Ambeed
Warning

A880402-10G A880402-1G A880402-25G A880402-5G A880402-250MG
76832-726EA 31.83 USD
76832-726 76832-728 76832-730 76832-732 77684-844
2,4-Difluoro-5-iodobenzoic acid 95%
2,4-Difluoro-5-iodobenzoic acid

Precaución: For research use only.

Formula: C₇H₃F₂IO₂
MW: 284 g/mol
MDL Number: MFCD11226506
CAS Number: 161531-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR