3,5-Dibromopyridin-4(1H)-one 98%

Supplier: Ambeed
Warning

A878174-1G A878174-5G
76832-722EA 26.53 USD
76832-722 76832-724
3,5-Dibromopyridin-4(1H)-one 98%
3,5-Dibromopyridin-4(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃Br₂NO
MW: 252.89 g/mol
MDL Number: MFCD00955615
CAS Number: 28419-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR