5-Nitropyrimidine-2,4-diamine 98%

Supplier: Ambeed
Warning

A876095-10G A876095-1G A876095-25G A876095-5G
76832-702EA 334 USD
76832-702 76832-704 76832-706 76832-708
5-Nitropyrimidine-2,4-diamine 98%
5-Nitropyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₄H₅N₅O₂
MDL Number: MFCD00056035
CAS Number: 18620-73-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR