4-(3,4-Dichlorophenyl)-4-oxobutanoic acid 95%

Supplier: Ambeed
Warning

A873627-100MG A873627-1G A873627-250MG
76832-678EA 33.95 USD
76832-678 76832-680 76832-682
4-(3,4-Dichlorophenyl)-4-oxobutanoic acid 95%
4-(3,4-Dichlorophenyl)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈Cl₂O₃
MW: 247.0747 g/mol
MDL Number: MFCD01464149
CAS Number: 50597-19-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR