2-Iodo-5-nitroaniline 97%

Supplier: Ambeed
Warning

A871769-100MG A871769-1G A871769-250MG
76832-672EA 27.59 USD
76832-672 76832-674 76832-676
2-Iodo-5-nitroaniline 97%
2-Iodo-5-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Number: MFCD16877042
CAS Number: 5459-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR