(11bR)-2,6-Bis(4-(tert-butyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A870344-1G
A870344-250MG
76832-656EA
802.05
USD
76832-656
76832-658
(11bR)-2,6-Bis(4-(tert-butyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide ≥98%, ee 99%
(11bR)-2,6-Bis(4-(tert-butyl)phenyl)-4-hydroxydinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepine 4-oxide
Precaución: for Research Use Only
Formula:
C₄₀H₃₇O₄P MW: 612.69 g/mol |
MDL Number:
MFCD27920532 CAS Number: 861909-30-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...