(1-Isobutyl-1H-pyrazol-5-yl)boronic acid 96%

Supplier: Ambeed
Warning

A870252-100MG A870252-1G A870252-250MG A870252-5G
76832-650EA 118 USD
76832-650 76832-652 76832-654 77886-870
(1-Isobutyl-1H-pyrazol-5-yl)boronic acid 96%
(1-Isobutyl-1H-pyrazol-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₃BN₂O₂
MW: 168.00 g/mol
MDL Number: MFCD12407379
CAS Number: 847818-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR