4-Chloro-2-methoxyquinoline 95%

Supplier: Ambeed
Warning

A870191-100MG A870191-250MG A870191-1G A870191-5G
76832-648EA 17 USD
76832-648 77595-404 77886-868 77886-869
4-Chloro-2-methoxyquinoline 95%
4-Chloro-2-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
MDL Number: MFCD18254961
CAS Number: 4295-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR