Picolinic anilide 95%

Supplier: Ambeed
Warning

Synonyms: N-Phenylpicolinamide

A868650-100MG A868650-1G A868650-250MG A868650-5G
76832-632EA 16 USD
76832-632 76832-634 76832-636 76832-638
Picolinic anilide 95%
Picolinic anilide

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O
MW: 198.2200 g/mol
MDL Number: MFCD00511984
CAS Number: 10354-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR