2-Bromo-6-chloro-3-methylpyridine 98%

Supplier: Ambeed
Warning

A867737-250MG A867737-1G
76832-622EA 60 USD
76832-622 77614-562
2-Bromo-6-chloro-3-methylpyridine 98%
2-Bromo-6-chloro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
MW: 206.46 g/mol
MDL Number: MFCD11044269
CAS Number: 867377-03-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR