1-Heptanesulfonic acid sodium salt monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: Sodium 1-heptanesulphonate monohydrate

A861530-5G A861530-100G A861530-25G
76832-618EA 6.37 USD
76832-618 77614-548 77684-823
1-Heptanesulfonic acid sodium salt monohydrate 98%
1-Heptanesulphonic acid sodium salt monohydrate

Precaución: For research use only.

Formula: C₇H₁₅NaO₃S·1H₂O
MW: 220.27 g/mol
MDL Number: MFCD00149550
CAS Number: 207300-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR