3-Isopropylbenzene-1-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A857684-1G A857684-250MG
76832-604EA 44 USD
76832-604 76832-606
3-Isopropylbenzene-1-sulfonyl chloride 95%
3-Isopropylbenzene-1-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂S
MDL Number: MFCD09730000
CAS Number: 71530-58-0
UN: 1760
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR