1-(2-Bromo-4,5-difluorophenyl)ethanone 95%

Supplier: Ambeed
Warning

A855650-250MG A855650-5G
76832-594EA 19.1 USD
76832-594 76832-596
1-(2-Bromo-4,5-difluorophenyl)ethanone 95%
1-(2-Bromo-4,5-difluorophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₅BrF₂O
MW: 235.02 g/mol
MDL Number: MFCD23135345
CAS Number: 127371-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR