5-Ethoxy-2-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: Dihydroxy(5-ethoxy-2-fluorophenyl)borane, (5-Ethoxy-2-fluorophenyl)boronic acid, 5-Ethoxy-2-fluorobenzeneboronic acid, (5-Ethoxy-2-fluorophenyl)metaboric acid

A852664-1G A852664-250MG A852664-5G
76832-582EA 20 USD
76832-582 76832-584 76832-586
5-Ethoxy-2-fluorophenylboronic acid 98%
5-Ethoxy-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MDL Number: MFCD05664321
CAS Number: 900174-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR