Pyrene-4,5-dione 95%

Supplier: Ambeed
Warning

A850914-100MG A850914-1G A850914-250MG
76832-576EA 150.65 USD
76832-576 76832-578 76832-580
Pyrene-4,5-dione 95%
Pyrene-4,5-dione

Precaución: for Research Use Only

Formula: C₁₆H₈O₂
MW: 232.23 g/mol
MDL Number: MFCD00089611
CAS Number: 6217-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR