2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid,pinacol ester, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid pinacol ester

A846351-1G A846351-250MG A846351-5G
76832-562EA 34 USD
76832-562 76832-564 76832-566
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester 97%
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BFO₄
MW: 280.0997 g/mol
MDL Number: MFCD11855962
CAS Number: 603122-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR