5,10,15,20-Tetrakis(4-fluorophenyl)-21H,23H-porphine 98%

Supplier: Ambeed
Warning

A845717-100MG A845717-1G A845717-250MG
76832-556EA 33 USD
76832-556 76832-558 76832-560
5,10,15,20-Tetrakis(4-fluorophenyl)-21H,23H-porphine 98%
5,10,15,20-Tetrakis(4-fluorophenyl)-21H,23H-porphine

Precaución: for Research Use Only

Formula: C₄₄H₂₆F₄N₄
MDL Number: MFCD01941197
CAS Number: 37095-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR